Alfa Aesar™ Zinc bromide hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1296014

Quantity 25g

 View more versions of this product

Catalog No. AA1296014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7699-45-8
    Formula Weight 225.19 (Anhydrous)
    MDL Number MFCD00011294
    UN Number UN3260
    Merck Index 14,10129
    Chemical Name or Material Zinc bromide hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 225.19 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    Health Hazard 1 Danger
    Physical Form Crystalline
    MDL Number MFCD00011294
    CAS 7699-45-8
    UN Number UN3260
    Merck Index 14,10129
    Quantity 25g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Zinc bromide hydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit