Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride, 0.7M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
7646-85-7,109-99-9
Cl2Zn
136.3
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
7646-85-7,109-99-9
Cl2Zn
136.3
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
Linear Formula ZnCl2
Merck Index 15, 10331
Solubility Information Solubility in water: miscible
Formula Weight 136.3
Physical Form Liquid
Concentration 0.65 to 0.8M (argentometry)
Density 0.9800g/mL
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: miscible.
Specific Gravity 0.98
Water 0.1% Max.
Boiling Point 66.0°C
Color Red
Quantity 100mL
Flash Point −22°C
Chemical Name or Material Zinc chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing cancer.
May cause respiratory irritation.
May form

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 231-592-