Learn More
Zinc chloride, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389458000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
66.0°C | |
Red | |
Liquid | |
Cl2Zn | |
MFCD00011295 | |
15, 10331 | |
Solubility in water: miscible | |
Cl[Zn]Cl | |
136.3 | |
CHEBI:49976 | |
0.65 to 0.8M (argentometry) | |
0.98 |
Zinc chloride | |
7646-85-7, 109-99-9 | |
800 mL | |
ZnCl2 | |
−22°C | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.3 | |
AcroSeal™ Glass Bottle | |
0.9800g/mL |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing cancer.
May cause respiratory irritation.
May form
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 231-592-
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only