Learn More
Alfa Aesar™ Zinc chloride, 1.9M in 2-MeTHF
Manufacturer:  Alfa Aesar™ H54398AE
Quantity 100mL
 View more versions of this product
Catalog No. AAH54398AE
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
7646-85-7
ZnCl2
MFCD00011295
14,10132
Cl2Zn
136.29
UN2924
Zinc chloride
Specifications
7646-85-7
ZnCl2
MFCD00011295
14,10132
100mL
Cl2Zn
136.29
UN2924
1.9M in 2-MeTHF
Zinc chloride
Safety and Handling
SDS & Safety Documents
GHS H Statement
H225-H314-H302
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : TSCA