Alfa Aesar™ Zinc chloride, 1M in diethyl ether, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 42781AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42781AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-85-7
    Linear Formula ZnCl2
    Formula Weight 136.29
    MDL Number MFCD00011295
    UN Number UN2924
    Merck Index 14,10132
    Chemical Name or Material Zinc chloride

    Specifications

    Formula Weight 136.29
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011295
    Health Hazard 2 GHS H Statement
    H225-H314-H302-H336
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.
    Color Colorless
    Quantity 500mL
    Concentration 1M in diethyl ether
    Sensitivity Hygroscopic
    Health Hazard 1 Danger
    TSCA TSCA
    Density 0.835g/mL
    Linear Formula ZnCl2
    CAS 7646-85-7
    UN Number UN2924
    Merck Index 14,10132
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 3; Packaging Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Packaging ChemSeal™ bottles
    Physical Form Liquid
    Chemical Name or Material Zinc chloride

    GHS H Statement
    H225-H314-H302-H336
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit