Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride, 98.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
7646-85-7
Cl2Zn
136.29
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
7646-85-7
Cl2Zn
136.29
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
Assay Percent Range 98% min. (Complexometry)
Linear Formula ZnCl2
Ammonium 50ppm max.
Fieser 01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391
Merck Index 15, 10331
Solubility Information Solubility in water: 432g/100mL (25°C). Other solubilities: soluble in alcohol,glycerol and ether,freely soluble in acetone
Formula Weight 136.29
Physical Form Crystalline Powder, Crystals or Chunks
Percent Purity 98.5%
Grade Analysis
Cadmium (Cd) 5ppm max.
Calcium (Ca) 10ppm max.
Copper (Cu) 10ppm max.
Density 0.84
Insoluble Matter 0.005% max
Iron (Fe) 5ppm max.
Lead (Pb) 10ppm max.
Magnesium (Mg) 100ppm max.
Nitrate 30ppm max.
Oxide Chloride (ZnO) 1.2% max.
Packaging Plastic bottle
Potassium (K) 200ppm max.
Sodium (Na) 50ppm max.
Solubility Solubility in water: 432 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and ether, freely soluble in acetone
Sulfate 20ppm max.
Total Nitrogen 0.002% max.
Boiling Point 732.0°C
Color White
Melting Point 293.0°C
Quantity 500g
Chemical Name or Material Zinc chloride, p.a.
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-592-0

RTECSNumber : ZH1400000

TSCA : TSCA