Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride, 99.99%, (trace metal basis), anhydrous, ACROS Organics™

Click to view available options
Quantity:
5g
25g
7646-85-7
Cl2Zn
136.29
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
7646-85-7
Cl2Zn
136.29
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula ZnCl2
Merck Index 15, 10331
Solubility Information Solubility in water: 432g/100mL (25°C). Other solubilities: soluble in alcohol,glycerol and ether,freely soluble in acetone
Formula Weight 136.29
Physical Form Free Flowing Small Crystals
Percent Purity 99.99%
Grade Trace Metal Basis
Density 0.95
Packaging Ampule packed in tin can
Solubility Solubility in water: 432 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and ether, freely soluble in acetone
Total Trace Metal Impurities 0.01% max
Boiling Point 732.0°C
Color White
Melting Point 293.0°C
Quantity 25g
Chemical Name or Material Zinc chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-592-