Zinc chloride, ACS, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar 12307A7

Quantity 5kg

 View more versions of this product

Catalog No. AA12307A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-85-7
    Physical Form Lumps
    Assay Percent Range 97%
    Molecular Formula Cl2Zn
    Formula Weight 136.28
    MDL Number MFCD00011295
    Chemical Name or Material Zinc chloride, For analysis ACS

    Specifications

    Formula Weight 136.28
    Quantity 5kg
    EINECS Number 231-592-0
    Physical Form Lumps
    Assay Percent Range 97%
    Molecular Formula Cl2Zn
    MDL Number MFCD00011295
    CAS 7646-85-7
    Chemical Name or Material Zinc chloride, For analysis ACS

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit