Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Crystalline Powder, USP, 97-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Amber Glass Bottle
7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc(2+) dichloride
[Cl-].[Cl-].[Zn++]
7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc(2+) dichloride
[Cl-].[Cl-].[Zn++]

Z1148, 7646-85-7, ZnCl2

Spectrum™ Zinc Chloride, Crystalline Powder, USP
Packaging Amber Glass Bottle
Quantity 2.5 kg
CAS Max % 1
Percent Purity 97 to 100.5%
Grade USP