Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
Formula Weight 136.3
Physical Form Granular
Packaging Poly Bottle
Specific Gravity 1L = 2.91kg
Quantity 500g
Grade Multi-Compendial/USP
Chemical Name or Material Zinc Chloride, Endotoxin Tested