Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Granular, USP, 97-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc(2+) dichloride
[Cl-].[Cl-].[Zn++]
7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc(2+) dichloride
[Cl-].[Cl-].[Zn++]

ZI105, 7646-85-7, ZnCl2

Spectrum™ Zinc Chloride, Granular, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 1
Percent Purity 97 to 100.5%
Grade USP