Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Reagent grade, ≥98%, Honeywell™

Click to view available options
Quantity:
5g
100g
500g
1kg
Packaging:
Poly Bottle
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
Linear Formula ZnCl2
UN Number UN2331
Formula Weight 136.3g/mol
Percent Purity ≥98%
Grade Reagent
Density 2.93g/cu.cm
Packaging Poly Bottle
Vapor Pressure 1mmHg (428°C)
Physical State Solid
Color White
Melting Point 293°C
Quantity 5g
Chemical Name or Material Zinc Chloride
Show More Show Less

P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H302-H314-H410

DOTInformation : 8/PGIII

EINECSNumber : 231-592-0

RTECSNumber : ZH1400000