Alfa Aesar™ Zinc cyanide, 85%

Electroplating, reagent

Manufacturer: Alfa Aesar™ 1415857

Quantity 4 x 500g

 View more versions of this product

Catalog No. AA1415857

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 557-21-1
    Formula Weight 117.42
    MDL Number MFCD00011292
    UN Number UN1713
    Beilstein 4124366
    Merck Index 14,10135
    Chemical Name or Material Zinc cyanide

    Specifications

    Formula Weight 117.42
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    MDL Number MFCD00011292
    Beilstein 4124366
    Color White
    RTECS Number ZH1575000
    Sensitivity Hygroscopic
    CAS 557-21-1
    UN Number UN1713
    Merck Index 14,10135
    Solubility Information Insoluble in water, alcohol; Soluble in solutions of alkali cyanides or hydroxides
    Health Hazard 1 Danger
    Density 1.852g/mL
    TSCA TSCA
    Melting Point 800°C (dec.)
    Quantity 4 x 500g
    Health Hazard 2 GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ZINC CYANIDE
    Chemical Name or Material Zinc cyanide
    Assay 85%

    GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit