Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc cyclohexanebutyrate dihydrate, AAS

Manufacturer:  Alfa Aesar™ 3973406

Quantity 5g

 View more versions of this product

Catalog No. AA3973406


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

38582-18-2
MFCD00035537
Zinc cyclohexanebutyrate dihydrate, AAS
439.91 (403.87 Anhydrous)
Cyclohexanebutyric acid zinc salt dihydrate; 4-Cyclohexylbutyric acid zinc salt dihydrate

Specifications

77° to 80°C
5g
439.91 (403.87 Anhydrous)
Cyclohexanebutyric acid zinc salt dihydrate; 4-Cyclohexylbutyric acid zinc salt dihydrate
38582-18-2
Powder
MFCD00035537
Zinc cyclohexanebutyrate dihydrate, AAS

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit