Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Zinc cyclohexanebutyrate dihydrate, AAS
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 1g |
Melting Point | 77°C to 80°C |
Formula Weight | 439.91 (403.87 Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Zinc cyclohexanebutyrate dihydrate, AAS |
Safety and Handling
EINECSNumber : 254-017-5
TSCA : Yes
Recommended Storage : Ambient temperatures