Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals

Catalog Number AA1869736
Click to view available options
Quantity:
500 g
2 kg
14324-55-1
C10H20N2S4Zn
361.904
MFCD00064798
RKQOSDAEEGPRER-UHFFFAOYSA-L
zinc diethyldithiocarbamate, ethyl ziram, zinc bis diethyldithiocarbamate, diethyldithiocarbamic acid zinc salt, zinc n,n-diethylcarbamodithioate, ethyl zimate, unii-icw4708z8g, ditiocarb zinc, zdc, zdec
26633
zinc;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]
14324-55-1
C10H20N2S4Zn
361.904
MFCD00064798
RKQOSDAEEGPRER-UHFFFAOYSA-L
zinc diethyldithiocarbamate, ethyl ziram, zinc bis diethyldithiocarbamate, diethyldithiocarbamic acid zinc salt, zinc n,n-diethylcarbamodithioate, ethyl zimate, unii-icw4708z8g, ditiocarb zinc, zdc, zdec
26633
zinc;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]

Chelating agent

Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 183°C
Color White
Density 1.48 g/mL
pH 7.8
Flash Point 204°C (400°F)
Odor Odorless
Linear Formula Zn[CS2N(CH2CH3)2]2
Quantity 500 g
UN Number UN3077
Solubility Information Insoluble in water. Soluble in benzene carbon disulfide and organic liquids.
Formula Weight 361.9
Concentration or Composition (by Analyte or Components) Zn 17 to 19.5%
Physical Form Crystalline
Chemical Name or Material Zinc diethyldithiocarbamate
Show More Show Less
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 238-270-9
RTECSNumber ZH0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit