Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc finger protein 833 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $349.40

Specifications

Antigen Zinc finger protein 833
Immunogen Synthetic peptide directed towards the N terminal of human LOC401898. Peptide sequence MVMHSEDEPYKCKFCGKAFDNLHLYLTHERTHTGEKPYECNKCGKAFSCS.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17936020
View Documents
Novus Biologicals
NBP17936020UL
20ul
Each for $152.22
Add to cart
 
NBP179360
View Documents
Novus Biologicals
NBP179360
100 ul
Each for $349.40
Add to cart
 
Description

Description

Zinc finger protein 833 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Zinc finger protein 833
Rabbit
Polyclonal
Unconjugated
401898
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the N terminal of human LOC401898. Peptide sequence MVMHSEDEPYKCKFCGKAFDNLHLYLTHERTHTGEKPYECNKCGKAFSCS.
Primary
IgG
zinc finger protein 833, pseudogene, ZNF833
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit