Zinc Flake, -325 mesh, 97% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4507030

Quantity 250g

 View more versions of this product

Catalog No. AA4507030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-66-6
    Physical Form Flakes
    Assay Percent Range 97% (Metals basis)
    MDL Number MFCD00011291
    Chemical Name or Material Zinc

    Specifications

    Quantity 250g
    Color Gray
    DOT Information DOT Class: 4.3, Packaging Group: II
    Density 7.14g/cm3
    EINECS Number 231-175-3
    Physical Form Flakes
    Assay Percent Range 97% (Metals basis)
    MDL Number MFCD00011291
    CAS 7440-66-6
    Odor Odorless
    Health Hazard 1 H250-H260-H400-H410: Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Appearance Gray flake
    Health Hazard 2 If swallowed seek medical advice immediately and show this container or label. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    Chemical Name or Material Zinc

    If swallowed seek medical advice immediately and show this container or label. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H250-H260-H400-H410: Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit