Zinc fluoride, anhydrous, 99% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1154118

Quantity 50g

 View more versions of this product

Catalog No. AA1154118

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7783-49-5
    Physical Form Powder
    Assay Percent Range 99% min. (anhydrous)
    Molecular Formula ZnF2
    Formula Weight 103.37
    MDL Number MFCD00011298

    Specifications

    Formula Weight 103.37
    Quantity 50g
    Color White
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99% min. (anhydrous)
    Molecular Formula ZnF2
    MDL Number MFCD00011298
    CAS 7783-49-5

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit