Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc foil, 0.5mm (0.02 in.) thick, Puratronic™, 99.994% (metals basis)

Click to view available options
Quantity:
50 x 50mm
100 x 100mm
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
Quantity 50 x 50mm
Weight ≈8.9g/50x50mm
Odor Odorless
Physical Form Foil
Percent Purity 99.994%
Grade Puratronic™
Chemical Name or Material Zinc foil, 0.5mm (0.02 in.) thick
Assay (metals basis)

EINECSNumber : 231-175-3

TSCA : Yes

Recommended Storage : Ambient temperatures