Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc formate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Zinc formate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc formate is used as a pharmaceutical intermediate.
Solubility
Soluble in water (52 g/L at 20°C).
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
Zinc formate is used as a pharmaceutical intermediate.
Solubility
Soluble in water (52 g/L at 20°C).
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Quantity | 100 g |
UN Number | UN3077 |
Solubility Information | Soluble in water (52g/L at 20°C). |
Formula Weight | 155.4 |
Percent Purity | 98% |
Chemical Name or Material | Zinc formate |
Safety and Handling
Hazard Category | H302-H319 |
Hazard Statement | GHS H Statement H302-H319 Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 209-176-5 |
RTECSNumber | LR0725000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction