Zinc formate dihydrate, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 13797A1

Quantity 1kg

 View more versions of this product

Catalog No. AA13797A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 5970-62-7
    Physical Form Powder
    Assay Percent Range 98%
    Molecular Formula Zn(CHO2)2·2H2O
    Formula Weight 191.43 (155.40 Anhydrous)
    MDL Number MFCD00050836
    Chemical Name or Material Zinc formate dihydrate

    Specifications

    Formula Weight 191.43 (155.40 Anhydrous)
    Quantity 1kg
    EINECS Number 209-176-5
    Physical Form Powder
    Assay Percent Range 98%
    Molecular Formula Zn(CHO2)2·2H2O
    MDL Number MFCD00050836
    CAS 5970-62-7
    Chemical Name or Material Zinc formate dihydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit