Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc Gluconate, FCC, 97-102%, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Zinc Gluconate, FCC is a popular form for the delivery of zinc as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS | 4468-02-4 |
CAS Max % | 1 |
Packaging | Poly Bottle |
Molecular Formula | C12H24O14Zn |
Quantity | 500 g |
InChI Key | ZSKINHGFQOKUDM-IYEMJOQQSA-N |
SMILES | [Zn].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O |
IUPAC Name | zinc gluconate |
Molecular Weight (g/mol) | 457.69 |
Percent Purity | 97 to 102% |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction