Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc (Granular, 20 Mesh/Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Poly Bottle
Specifications
Color | Blue |
Melting Point | 419°C |
Boiling Point | 907°C |
Atomic Weight | 65.37 |
Density | 7.14g/cm³ |
Assay Percent Range | ≥99.8 % |
Quantity | 500 g |
Identification | Pass Test |
Formula Weight | 65.41 |
Percent Purity | ≥99.8% |
Show More |
Safety and Handling
Hazard Statement | CAUTION! |
Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction