Alfa Aesar™ Zinc iodide, 98%

Manufacturer: Alfa Aesar™ 1166130

Quantity 250g

 View more versions of this product

Catalog No. AA1166130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10139-47-6
    Linear Formula ZnI2
    Formula Weight 319.18
    MDL Number MFCD00011299
    UN Number UN3260
    Merck Index 14,10140
    Chemical Name or Material Zinc iodide

    Specifications

    Formula Weight 319.18
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 625°C (dec.)
    Melting Point 446°C
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Sensitivity Hygroscopic
    MDL Number MFCD00011299
    CAS 10139-47-6
    Merck Index 14,10140
    Quantity 250g
    Solubility Information Freely soluble in water, glycerol, alcohol, ether
    Density 4.74g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline
    Linear Formula ZnI2
    UN Number UN3260
    Health Hazard 1 Danger
    Color White to pale yellow
    Chemical Name or Material Zinc iodide
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit