Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Iodide, Purified, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
10139-47-6
I2Zn
319.19
UAYWVJHJZHQCIE-UHFFFAOYSA-L
zinc(2+) diiodide
[Zn++].[I-].[I-]
10139-47-6
I2Zn
319.19
UAYWVJHJZHQCIE-UHFFFAOYSA-L
zinc(2+) diiodide
[Zn++].[I-].[I-]

Z1073, 10139-47-6, ZnI2

Spectrum™ Zinc Iodide, Purified is used in industrial radiography as an x-ray opaque penetrant and as a stain in electron microscopy. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Grade Purified
CAS Max % 1
Percent Purity 98%
Packaging Amber Glass Bottle
Quantity 100 g