Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc, Mossy, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7440-66-6
Zn
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
7440-66-6
Zn
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 7.14kg
Quantity 500g
Packaging Poly Bottle
Formula Weight 65.39
Physical Form Mossy
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Zinc