Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Zinc naphthenate, ≈67% in mineral spirits (10% Zn)
Click to view available options
Quantity:
500g
2kg
Specifications
Color | Colorless |
Flash Point | >37.8° (100°F) |
Quantity | 2kg |
Density | 1.01 g/mL |
UN Number | UN1993 |
Physical Form | Liquid |
Chemical Name or Material | Zinc naphthenate |
Safety and Handling
GHS H Statement
H350-H304-H226-H315-H319-H335
May cause cancer.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P332+P313-P363-P370+P378q-P501c
H226-H304-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures