Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals

Catalog Number AA41312A7
Click to view available options
Quantity:
250 g
1 kg
5 kg
27253-29-8
C20H40O4Zn
409.916
MFCD00014384
VADFQXONMAETJP-UHFFFAOYSA-N
zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
117064711
7,7-dimethyloctanoic acid;zinc
CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]
27253-29-8
C20H40O4Zn
409.916
MFCD00014384
VADFQXONMAETJP-UHFFFAOYSA-N
zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
117064711
7,7-dimethyloctanoic acid;zinc
CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]

Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Flash Point 102°C (216°F)
Quantity 5 kg
Solubility Information Immiscible with water.
Formula Weight 407.89
Physical Form Liquid
Chemical Name or Material Zinc neodecanoate
EINECSNumber 248-370-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit