Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Zinc Nitrate Hexahydrate, 98%, Reagent grade, Honeywell™

Click to view available options
Quantity:
100g
500g
7kg
Packaging:
Drum with inliner
Poly Bottle
10196-18-6
H12N2O12Zn
297.478
MFCD00149889
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc nitrate hexahydrate,zinc ii nitrte,zincnitratehexahydrate,acmc-1bzdd,n2o6zn.6h2o,ksc176g7t,zinc nitrate hexahydrate, puratronic,zinc nitrate,zinc 2+ hexahydrate dinitrate,zinc nitrate hexahydrate, p.a
15865313
zinc;dinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Zn+2]
10196-18-6
H12N2O12Zn
297.478
MFCD00149889
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc nitrate hexahydrate,zinc ii nitrte,zincnitratehexahydrate,acmc-1bzdd,n2o6zn.6h2o,ksc176g7t,zinc nitrate hexahydrate, puratronic,zinc nitrate,zinc 2+ hexahydrate dinitrate,zinc nitrate hexahydrate, p.a
15865313
zinc;dinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Zn+2]

Reagent Grade, 98%

  • ≥95% for general laboratory use.
Linear Formula Zn(NO3)2 · 6H2O
UN Number UN1514
Appearance Powder or Crystals or Flakes
Formula Weight 297.49g/mol
Percent Purity 98%
Grade Reagent
Density 2.060g/cm3
Packaging Poly Bottle
Solubility Soluble in water
Color White
Melting Point 38.9°C
Quantity 100g
Chemical Name or Material Zinc Nitrate Hexahydrate
Show More Show Less

P220-P261-P305 + P351 + P338

H272-H302-H315-H319-H335

DOTInformation : 5.1/PGII

EINECSNumber : 231-943-8

RTECSNumber : ZH4775000