Learn More
Zinc oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Zinc oxide is used as a sun screener. It serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. Zinc oxide plays an important role in textiles as UV-absorber. It can protect rubber from fungi and UV light. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1975°C |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.999%, metal basis |
Solubility Information | Insoluble in water. Soluble in dilute acetic acid,and dilute mineral acids. |
Formula Weight | 81.37 |
Density | 5.606 |
Chemical Name or Material | Zinc Oxide |
Safety and Handling
Hazard Category | Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. |
Hazard Statement | This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets |
DOTInformation | DOT Class: 9; Packaging Group: III |
EINECSNumber | 215-222-5 |
RTECSNumber | ZH4810000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.