Zinc oxide, 99.9995% (metals basis), Puratronic™, Alfa Aesar™

Manufacturer: Alfa Aesar 1285522

Quantity 100g

 View more versions of this product

Catalog No. AA1285522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-13-2
    Physical Form Powder
    Assay Percent Range 99.9995% metal basis
    Molecular Formula ZnO
    Formula Weight 81.37
    MDL Number MFCD00011300

    Specifications

    Formula Weight 81.37
    Quantity 100g
    Color White
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.9995% metal basis
    Molecular Formula ZnO
    MDL Number MFCD00011300
    CAS 1314-13-2

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit