Learn More
Alfa Aesar™ Zinc oxide based sulfur removal material, HiFUEL™ A310
Manufacturer:  Alfa Aesar™ 4546936
Quantity 500g
 View more versions of this product
Catalog No. AA4546936
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00011300
Zinc oxide based sulfur removal material HiFUEL™ A310
Specifications
Spheres
500g
Zinc oxide based sulfur removal material HiFUEL™ A310
MFCD00011300
2 to 5mm dia.
Safety and Handling
SDS & Safety Documents
TSCA : TSCA