Learn More
Zinc oxide, NanoArc™ ZN-0605, Thermo Scientific Chemicals
Description
Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis. Used in electronic industry, instrumental industry, manufacture of electrical device, image recorder, rheostat, phosphor. Used as a sun screening agent in cosmetics, antibacterial and health protection antiager; UV protection; Piezoelectrics; High-temperature lubricant in gas turbine engines; Flame retardant; Adsorption; Dental cements; Environmental remediation; Gas sensors; Photocatalytic decontamination; Attenuation of ultraviolet light; Demilitarization of chemical and biological warfare agents; Cosmetics and cosmeceuticals; Electrodes for solar cells; Varistors; Pigments for paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1975°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | ZnO |
UN Number | UN3077 |
Merck Index | 14,10147 |
Solubility Information | Insoluble in water. |
Formula Weight | 81.37 |
Odor | Odorless |
Show More |
Safety and Handling
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 215-222-5 |
RTECSNumber | ZH4810000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.