Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2650, 50% in H2O, colloidal dispersion with nonionic dispersant

Manufacturer: Alfa Aesar™ 4490522

Quantity 100g

 View more versions of this product

Catalog No. AA4490522

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1314-13-2
    Molecular Formula ZnO
    Formula Weight 81.37
    MDL Number MFCD00011300
    UN Number UN3082
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2650, Colloidal dispersion with nonionic dispersant

    Specifications

    Physical Form Dry Powder (Typical)
    Formula Weight 81.37
    TSCA TSCA
    Molecular Formula ZnO
    CAS 1314-13-2
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Quantity 100g
    Odor Odorless
    Particle Size 70nm APS
    MDL Number MFCD00011300
    UN Number UN3082
    Concentration 50% in H2O
    Color White to off-white
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2650, Colloidal dispersion with nonionic dispersant

    Description

    • NanoShield™ ZN-3010 is a dispersion of zinc oxide in water designed to be incorporated in coatings applied to a fabric or non-woven substrates
    • Zinc oxide is generally used to provide UVa and UVb blocking

    See Product Certificates
  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit