Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2656, 50% in H2O, colloidal dispersion with nonionic dispersant and rheology modifier

Manufacturer: Alfa Aesar™ 4722236

Quantity 500g

 View more versions of this product

Catalog No. AA4722236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-13-2
    Formula Weight 81.37
    MDL Number MFCD00011300
    UN Number UN3082
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier

    Specifications

    Formula Weight 81.37
    MDL Number MFCD00011300
    Physical Form Powder
    CAS 1314-13-2
    Odor Odorless
    UN Number UN3082
    Concentration 50% in H2O
    Particle Size 70nm APS for dry (typical)
    DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Quantity 500g
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit