Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2656, 50% in H2O, colloidal dispersion with nonionic dispersant and rheology modifier

Manufacturer: Alfa Aesar™ 4722222

Quantity 100g

 View more versions of this product

Catalog No. AA4722222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-13-2
    Formula Weight 81.37
    MDL Number MFCD00011300
    UN Number UN3082
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier

    Specifications

    Formula Weight 81.37
    MDL Number MFCD00011300
    Quantity 100g
    CAS 1314-13-2
    UN Number UN3082
    Particle Size 70nm APS for dry (typical)
    Physical Form Powder
    Odor Odorless
    Concentration 50% in H2O
    DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit