Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc oxide, NanoShield™ ZN-2001, 50% in H2O, colloidal dispersion with dispersant and rheology modifier

Manufacturer:  Alfa Aesar™ 4500636

Quantity 500g

 View more versions of this product

Catalog No. AA4500636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1314-13-2
81.37
Zinc oxide, NanoShield™ ZN-2001, Colloidal dispersion with dispersant and rheology modifier
ZnO
MFCD00011300

Specifications

1314-13-2
ZnO
MFCD00011300
Odorless
500g
Dry Powder (Typical)
81.37
50% in H2O
70nm APS
Zinc oxide, NanoShield™ ZN-2001, Colloidal dispersion with dispersant and rheology modifier

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit