Alfa Aesar™ Zinc oxide, NanoShield™ ZN-2001, 50% in H2O, colloidal dispersion with dispersant and rheology modifier

Manufacturer: Alfa Aesar™ 45006-22

Quantity 100g

 View more versions of this product

Catalog No. AA4500622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1314-13-2
    Molecular Formula ZnO
    Formula Weight 81.37
    MDL Number MFCD00011300
    Chemical Name or Material Zinc oxide, NanoShield™ ZN-2001, Colloidal dispersion with dispersant and rheology modifier

    Specifications

    Physical Form Dry Powder (Typical)
    Formula Weight 81.37
    TSCA TSCA
    Particle Size 70nm APS
    Molecular Formula ZnO
    Quantity 100g
    Odor Odorless
    MDL Number MFCD00011300
    CAS 1314-13-2
    Concentration 50% in H2O
    Chemical Name or Material Zinc oxide, NanoShield™ ZN-2001, Colloidal dispersion with dispersant and rheology modifier

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit