Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc oxide, NanoTek™ C1, 99%, organosilane coated, hydrophobic, nonpolar, Thermo Scientific™
Click to view available options
Quantity:
500g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- NanoTek™ zinc oxide C1 has a hydrophobic, nonpolar organosilane coating to aid dispersing in organic systems
Specifications
CAS | 1314-13-2 |
Quantity | 500g |
Molecular Formula | OZn |
MDL Number | MFCD00011300 |
UN Number | UN3077 |
Merck Index | 14,10147 |
Synonym | zinc oxide, zinc white, amalox, permanent white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 |
InChI Key | XLOMVQKBTHCTTD-UHFFFAOYSA-N |
SMILES | O=[Zn] |
IUPAC Name | oxozinc |
Show More |
Safety and Handling
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
RTECSNumber | ZH4810000 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction