Alfa Aesar™ Zinc oxide, NanoTek™ C2, 99%, organosilane coated, hydrophobic, polar

Manufacturer: Alfa Aesar™ 4490122

Quantity 100g

 View more versions of this product

Catalog No. AA4490122

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1314-13-2
    Molecular Formula ZnO
    Formula Weight 81.37
    MDL Number MFCD00011300
    UN Number UN3077
    Merck Index 14,10147
    Chemical Name or Material Zinc oxide, NanoTek™ C2, Organosilane coated, Hydrophobic, Polar

    Specifications

    Formula Weight 81.37
    Odor Odorless
    Physical Form Powder
    Molecular Formula ZnO
    Surface Area 10 to 25m2/g
    MDL Number MFCD00011300
    CAS 1314-13-2
    Quantity 100g
    Particle Size 40 to 100nm APS
    UN Number UN3077
    Merck Index 14,10147
    Melting Point 1975°C
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number ZH4810000
    TSCA TSCA
    Chemical Name or Material Zinc oxide, NanoTek™ C2, Organosilane coated, Hydrophobic, Polar
    Assay 99%

    Description

    • NanoTek™ zinc oxide C2 has an oleophilic, polar coating to aid dispersing in organic systems

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit