Alfa Aesar™ Zinc oxide, NanoTek™ ZN-2551, 50% in H2O, colloidal dispersion with dispersant

Manufacturer: Alfa Aesar™ 4558836

Quantity 500g

 View more versions of this product

Catalog No. AA4558836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-13-2
    Molecular Formula OZn
    Linear Formula ZnO
    Formula Weight 81.37g/mol
    MDL Number MFCD00011300
    UN Number UN3082
    Chemical Name or Material Zinc oxide, NanoTek™ ZN-2551, Colloidal dispersion with dispersant

    Specifications

    Concentration 50% in H2O
    Odor Odorless
    Particle Size 50nm APS
    Color White
    Quantity 500g
    CAS 1314-13-2
    Physical Form Dry powder (typical)
    Molecular Formula OZn
    Linear Formula ZnO
    Formula Weight 81.37g/mol
    MDL Number MFCD00011300
    UN Number UN3082
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Chemical Name or Material Zinc oxide, NanoTek™ ZN-2551, Colloidal dispersion with dispersant

    Description

    • Water borne zinc oxide dispersion suitable for incorporation in water compatible coatings and formulations

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit