Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc oxide, NanoTek™ ZN-2610, 50% in mineral spirits, colloidal dispersion with dispersant

Manufacturer:  Alfa Aesar™ 4580136

Quantity 500g

 View more versions of this product

Catalog No. AA4580136

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Zinc oxide dispersed in mineral spirits suitable for incorporation in a wide variety of non-polar organic based coatings and formulations
Description & Specifications

Chemical Identifiers

1314-13-2
ZnO
MFCD00011300
Zinc oxide, NanoTek™ ZN-2610, Colloidal dispersion with dispersant
OZn
81.37
UN1993

Specifications

1314-13-2
OZn
81.37
UN1993
Ester-like
White
Zinc oxide, NanoTek™ ZN-2610, Colloidal dispersion with dispersant
Dry Powder (Typical)
ZnO
MFCD00011300
50% in mineral spirits
60nm APS
500g

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P273-P501a
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit