Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Oxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle
1314-13-2
OZn
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide,zinc white,amalox,permanent white,chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
1314-13-2
OZn
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide,zinc white,amalox,permanent white,chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 81.39
Physical Form Powder
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 5.61kg
Quantity 500g
Chemical Name or Material Zinc Oxide