Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Oxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle

Chemical Identifiers

1314-13-2
OZn
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
CHEBI:36560

High quality chemicals for laboratory and specialized industrial use

SMILES O=[Zn]
IUPAC Name oxozinc
Formula Weight 81.39
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 5.61kg
Quantity 2kg
Chemical Name or Material Zinc Oxide
Grade Baker Analyzed™ ACS Reagent