Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Zinc Oxide, ReagentPlus™, 99.9%, Honeywell™

Click to view available options
Quantity:
100g
1kg
5kg
Packaging:
Drum with Inliner
Poly Bottle
1314-13-2
OZn
81.379
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide,zinc white,amalox,permanent white,chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
1314-13-2
OZn
81.379
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide,zinc white,amalox,permanent white,chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]

Reagent Grade, powder, <5 μm particle size, 99.9%

  • ≥95% for general laboratory use.
Linear Formula ZnO
UN Number UN3077
Formula Weight 81.39g/mol
Physical Form Powder
Percent Purity 99.90%
Grade ReagentPlus™
Density 5.6g/cu.cm
Packaging Poly Bottle
Particle Size <5μm
Color White
Melting Point 1975°C
Quantity 100g
Chemical Name or Material Zinc Oxide
Show More Show Less

P273-P501

H410

DOTInformation : 9/PGIII

EINECSNumber : 215-222-5

RTECSNumber : ZH4810000