Alfa Aesar™ Zinc phosphide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4545904

Quantity 2g

 View more versions of this product

Catalog No. AA4545904

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-84-7
    Molecular Formula Zn3P2
    Formula Weight 258.1
    MDL Number MFCD00049631
    UN Number UN1714
    Merck Index 14,10152
    Chemical Name or Material Zinc phosphide

    Specifications

    Formula Weight 258.1
    Health Hazard 3 GHS P Statement
    P231+P232-P280-P233-P301+P310-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Particle Size 6mm & down
    MDL Number MFCD00049631
    Quantity 2g
    Density 4.55g/mL
    Physical Form Pieces
    Boiling Point 1100°C
    Melting Point 420°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H260-H300
    In contact with water releases flammable gases which may ignite spontaneously.
    Fatal if swallowed.
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ZINC PHOSPHIDE
    TSCA TSCA
    Sensitivity Moisture Sensitive
    Molecular Formula Zn3P2
    CAS 1314-84-7
    UN Number UN1714
    Merck Index 14,10152
    RTECS Number ZH4900000
    Chemical Name or Material Zinc phosphide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H260-H300
    In contact with water releases flammable gases which may ignite spontaneously.
    Fatal if swallowed.

    GHS P Statement
    P231+P232-P280-P233-P301+P310-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit