Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Phthalocyanine 95.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
10g
25g
14320-04-8
C32H16N8Zn
577.916
MFCD00041955
PODBBOVVOGJETB-UHFFFAOYSA-N
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
2735172
CHEBI:51218
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]
14320-04-8
C32H16N8Zn
577.916
MFCD00041955
PODBBOVVOGJETB-UHFFFAOYSA-N
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
2735172
CHEBI:51218
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]
Color Blue-Purple
Quantity 10g
Formula Weight 577.92
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (T)
Chemical Name or Material Zinc Phthalocyanine