Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc, plasma standard solution, Specpure™, Zn 10,000µg/mL

Manufacturer:  Alfa Aesar™ 14404AD

Quantity 50mL

 View more versions of this product

Catalog No. AA14404AD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00011291
UN3264
Zinc plasma standard solution, Zn 10,000μg/mL
Specpure™

Specifications

Liquid
MFCD00011291
50mL
Specpure™
Zn in 5% HNO3
UN3264
Zinc plasma standard solution, Zn 10,000μg/mL

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit