Alfa Aesar™ Zinc propionate, 97%

Manufacturer: Alfa Aesar™ B2363618

Quantity 50g

 View more versions of this product

Catalog No. AAB2363618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 557-28-8
    Molecular Formula (CH3CH2CO2)2Zn
    Formula Weight 211.53
    MDL Number MFCD00045937
    Merck Index 14,10153
    Synonym Propionic acid zinc salt
    Chemical Name or Material Zinc propionate

    Specifications

    Formula Weight 211.53
    TSCA TSCA
    Molecular Formula (CH3CH2CO2)2Zn
    MDL Number MFCD00045937
    Quantity 50g
    CAS 557-28-8
    Merck Index 14,10153
    Synonym Propionic acid zinc salt
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Zinc propionate
    Assay 97%

    Description

    Applications
    Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

    Solubility
    Soluble in alcohol, ether, chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.

    Notes
    Hygroscopic, protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit